4'-Chlorobiphenyl-4-ylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
C3614-1G
C3614-5G
TCC3614-1GEA
82
USD
TCC3614-1G
TCC3614-5G
4'-Chlorobiphenyl-4-ylboronic acid (contains varying amounts of Anhydride)
4'-Chlorobiphenyl-4-ylboronic acid
Formula:
C₁₂H₁₀BClO₂ |
MDL Number:
MFCD08544389 CAS Number: 364044-44-0 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | 98.0 to 107.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...