m-β-Hydroxyethylsulfonyl aniline ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 2-[(3-Aminophenyl)sulphonyl]ethanol

A3204-1G A3204-5G
TCA3204-1GEA 139 USD
TCA3204-1G TCA3204-5G
m-β-Hydroxyethylsulfonyl aniline ≥98.0% (by titrimetric analysis)
m-β-Hydroxyethylsulphonyl aniline
Formula: C₈H₁₁NO₃S
MDL Number: MFCD00044070
CAS Number: 5246-57-1
EINECS: 226-046-3

Order Now

Specification Test Results

Appearance White to Brown powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 73.0 to 77.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR