3-(Chlorosulfonyl)benzoic acid ≥96.0% (by titrimetric analysis)

Supplier: TCI America
Danger

C3466-25G C3466-5G
TCC3466-25GEA 113 USD
TCC3466-25G TCC3466-5G
3-(Chlorosulfonyl)benzoic acid ≥96.0% (by titrimetric analysis)
3-(Chlorosulphonyl)benzoic acid
Formula: C₇H₅ClO₄S
MW: 220.63 g/mol
MDL Number: MFCD00024877
CAS Number: 4025-64-3
EINECS: 223-692-8
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (Argentmetric Titration) min. 96.0 %
Melting point 132.0 to 137.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR