1-Boc-4-bromopiperidine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: tert-Butyl 4-Bromopiperidine-1-carboxylate, 4-Bromopiperidine-1-carboxylic acid tert-Butyl ester, 1-Boc-4-bromopiperidine, 1-tert-Butoxycarbonyl-4-bromopiperidine, N-Boc-4-Bromopiperidine

B5585-1G B5585-5G
TCB5585-1GEA 47 USD
TCB5585-1G TCB5585-5G
1-Boc-4-bromopiperidine ≥98.0% (by GC, titration analysis)
1-Boc-4-bromopiperidine
Formula: C₁₀H₁₈BrNO₂
MW: 264.1594 g/mol
MDL Number: MFCD04115039
CAS Number: 180695-79-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 40.0 to 44.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR