3-Chloro-5-(trifluoromethyl)picolinic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 3-Chloro-5-(trifluoromethyl)-2-pyridinecarboxylic acid, 3-Chloro-5-(trifluoromethyl)pyridine-2-carboxylic acid, 3-Chloro-5-(trifluoromethyl)picolinic acid

C3524-1G C3524-5G
TCC3524-1GEA 48 USD
TCC3524-1G TCC3524-5G
3-Chloro-5-(trifluoromethyl)picolinic acid ≥98.0% (by GC, titration analysis)
3-Chloro-5-(trifluoromethyl)picolinic acid
Formula: C₇H₃ClF₃NO₂
MW: 225.60 g/mol
MDL Number: MFCD00277482
CAS Number: 80194-68-9

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR