4-Cyano-4'-hexylbiphenyl ≥98.0% (by GC)

Supplier: TCI America
Warning

C3154-1G C3154-5G
TCC3154-1GEA 109 USD
TCC3154-1G TCC3154-5G
4-Cyano-4'-hexylbiphenyl ≥98.0% (by GC)
4-Cyano-4'-hexylbiphenyl
Formula: C₁₉H₂₁N
Boiling Pt: 185 °C
Melting Pt: 14 °C
MDL Number: MFCD00075144
CAS Number: 41122-70-7
EINECS: 255-228-5
UN: 3082
ADR: 9,III

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid to cloudy liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR