5-Chlorobenzoxazole ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
C3509-5G
C3509-25G
TCC3509-5GEA
61
USD
TCC3509-5G
TCC3509-25G
5-Chlorobenzoxazole ≥98.0% (by GC)
5-Chlorobenzoxazole
Formula:
C₇H₄ClNO MW: 153.57 g/mol Boiling Pt: 114 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00216939 CAS Number: 17200-29-2 |
Specification Test Results
Appearance | White to Orange to Green powder to lump |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...