5-Chloro-2-methylbenzonitrile ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
C3339-1G
C3339-5G
TCC3339-1GEA
26
USD
TCC3339-1G
TCC3339-5G
5-Chloro-2-methylbenzonitrile ≥98.0% (by GC)
5-Chloro-2-methylbenzonitrile
Formula:
C₈H₆ClN MW: 151.5929 g/mol |
MDL Number:
MFCD00045599 CAS Number: 50712-70-4 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 43.0 to 47.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...