3-Amino-2,5-dibromopyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2,5-Dibromopyridin-3-amine

A3048-1G A3048-5G
TCA3048-1GEA 62 USD
TCA3048-1G TCA3048-5G
3-Amino-2,5-dibromopyridine ≥98.0% (by GC)
3-Amino-2,5-dibromopyridine
Formula: C₅H₄Br₂N₂
MW: 251.9100 g/mol
MDL Number: MFCD09266224
CAS Number: 90902-84-4

Order Now

Specification Test Results

Appearance White to Brown powder to crystal
Purity (GC) min. 98.0 %
Melting point 152.0 to 156.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR