1-Cyclopentenylboronic acid pinacol ester ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 2-(Cyclopent-1-en-1-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)cyclopentene, 1-Cyclopenteneboronic acid Pinacol ester, 2-(1-Cyclopentenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

C3385-1G C3385-5G
TCC3385-1GEA 45.11 USD
TCC3385-1G TCC3385-5G
1-Cyclopentenylboronic acid pinacol ester ≥98.0% (by GC)
1-Cyclopentenylboronic acid pinacol ester
Formula: C₁₁H₁₉BO₂
MW: 194.0784 g/mol
MDL Number: MFCD08669391
CAS Number: 287944-10-9

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR