3-Bromo-N-methylbenzamide ≥98.0% (by GC)
Supplier: TCI America
B5644-1G
B5644-5G
TCB5644-1GEA
72
USD
TCB5644-1G
TCB5644-5G
3-Bromo-N-methylbenzamide ≥98.0% (by GC)
3-Bromo-N-methylbenzamide
Formula:
C₈H₈BrNO |
MDL Number:
MFCD01060879 CAS Number: 49834-22-2 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 91.0 to 95.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...