4-(Dimethylcarbamoyl)phenylboronic acid ≥97.0% (by titrimetric analysis) (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: (4-(Dimethylcarbamoyl)phenyl)boronic acid, 4-(Dimethylcarbamoyl)benzeneboronic acid, 4-(N,N-Dimethylaminocarbonyl)phenylboronic acid

D5226-1G D5226-5G
TCD5226-1GEA 71 USD
TCD5226-1G TCD5226-5G
4-(Dimethylcarbamoyl)phenylboronic acid ≥97.0% (by titrimetric analysis) (contains varying amounts of Anhydride)
4-(Dimethylcarbamoyl)phenylboronic acid
Formula: C₉H₁₂BNO₃
MW: 193.01 g/mol
MDL Number: MFCD02258943
CAS Number: 405520-68-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR