5-Bromobenzoxazole ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 5-Bromobenzo[d]oxazole

B5515-1G B5515-5G
TCB5515-1GEA 69 USD
TCB5515-1G TCB5515-5G
5-Bromobenzoxazole ≥98.0% (by GC)
5-Bromobenzoxazole
Formula: C₇H₄BrNO
MW: 198.0200 g/mol
MDL Number: MFCD03095026
CAS Number: 132244-31-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 36.0 to 40.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR