4,5-Dibromo-3(2H)-pyridazinone ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4,5-Dibromopyridazin-3(2H)-one

D5166-1G D5166-5G
TCD5166-1GEA 45 USD
TCD5166-1G TCD5166-5G
4,5-Dibromo-3(2H)-pyridazinone ≥98.0% (by GC, titration analysis)
4,5-Dibromo-3(2H)-pyridazinone
Formula: C₄H₂Br₂N₂O
MDL Number: MFCD00023641
CAS Number: 5788-58-9

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR