4-Chloro-2-ethoxyphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 4-Chloro-2-ethoxybenzeneboronic acid

C3267-1G C3267-200MG
TCC3267-1GEA 88 USD
TCC3267-1G TCC3267-200MG
4-Chloro-2-ethoxyphenylboronic acid (contains varying amounts of Anhydride)
4-Chloro-2-ethoxyphenylboronic acid
Formula: C₈H₁₀BClO₃
MW: 200.43 g/mol
MDL Number: MFCD03094940
CAS Number: 850568-80-8

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR