2-Bromo-3-fluorophenol ≥98.0% (by GC)

Supplier: TCI America
Warning

B5224-5G B5224-1G
TCB5224-5GEA 176 USD
TCB5224-5G TCB5224-1G
2-Bromo-3-fluorophenol ≥98.0% (by GC)
2-Bromo-3-fluorophenol
Formula: C₆H₄BrFO
MW: 190.9978 g/mol
Boiling Pt: 123 °C
MDL Number: MFCD04037220
CAS Number: 443-81-2

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR