(2-Carboxyethyl)triphenylphosphonium bromide ≥96.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

C3309-25G C3309-5G
TCC3309-25GEA 154 USD
TCC3309-25G TCC3309-5G
(2-Carboxyethyl)triphenylphosphonium bromide ≥96.0% (by HPLC, titration analysis)
(2-Carboxyethyl)triphenylphosphonium bromide
Formula: C₂₁H₂₀BrO₂P
MW: 415.2601 g/mol
MDL Number: MFCD00031698
CAS Number: 51114-94-4

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 96.0 area%
Purity (Neutralization titration) min. 96.0 %
Melting point 195.0 to 199.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR