2-Amino-4-fluoropyridine ≥98.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
A2976-1G
A2976-5G
TCA2976-1GEA
122
USD
TCA2976-1G
TCA2976-5G
2-Amino-4-fluoropyridine ≥98.0% (by GC, titration analysis)
2-Amino-4-fluoropyridine
Formula:
C₅H₅FN₂ |
MDL Number:
MFCD09260906 CAS Number: 944401-77-8 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 100.0 to 104.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...