4'-Benzyloxy-2-bromopropiophenone ≥98.0% (by GC)

Supplier: TCI America
Warning

B5990-25G B5990-5G
TCB5990-25GEA 154 USD
TCB5990-25G TCB5990-5G
4'-Benzyloxy-2-bromopropiophenone ≥98.0% (by GC)
4'-Benzyloxy-2-bromopropiophenone
Formula: C₁₆H₁₅BrO₂
MDL Number: MFCD00017862
CAS Number: 35081-45-9
EINECS: 252-351-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 77.0 to 81.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR