2-Amino-5-bromobenzophenone ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

A3311-1G A3311-5G
TCA3311-1GEA 94 USD
TCA3311-1G TCA3311-5G
2-Amino-5-bromobenzophenone ≥98.0% (by GC, titration analysis)
2-Amino-5-bromobenzophenone
Formula: C₁₃H₁₀BrNO
MDL Number: MFCD00059962
CAS Number: 39859-36-4

Order Now

Specification Test Results

Appearance Light yellow to Amber to Dark green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 111.0 to 115.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR