2-Amino-4'-bromobenzophenone ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

A3162-1G A3162-5G
TCA3162-1GEA 46 USD
TCA3162-1G TCA3162-5G
2-Amino-4'-bromobenzophenone ≥98.0% (by titrimetric analysis)
2-Amino-4'-bromobenzophenone
Formula: C₁₃H₁₀BrNO
MDL Number: MFCD02181034
CAS Number: 1140-17-6

Order Now

Specification Test Results

Appearance Light orange to Yellow to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 109.0 to 113.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR