1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane ≥94.0% (by GC)
Supplier: TCI America
|
Danger
|
Synonyms:
Chlorodimethyl(3,3,4,4,5,5,6,6,7,7,8,8,8-tridecafluoro-n-octyl)silane
C1857-1G
C1857-5G
TCC1857-1GEA
60
USD
TCC1857-1G
TCC1857-5G
1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane ≥94.0% (by GC)
1H,1H,2H,2H-Perfluorooctyldimethylchlorosilane
Formula:
C₁₀H₁₀ClF₁₃Si MW: 440.71 Boiling Pt: 80 °C Flash Pt: 52 °C |
MDL Number:
MFCD00042359 CAS Number: 102488-47-1 UN: 2986 ADR: 8,II |
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 94.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...