5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carboxaldehyde
C3559-1G
C3559-5G
TCC3559-1GEA
66
USD
TCC3559-1G
TCC3559-5G
5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde ≥98.0% (by GC)
5-Chloro-3-methyl-1-phenyl-1H-pyrazole-4-carbaldehyde
Formula:
C₉H₁₁ClN₂O MW: 220.65 g/mol |
MDL Number:
MFCD00100755 CAS Number: 947-95-5 |
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 143.0 to 148.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...