4'-Acetoxy-2'-hydroxyacetophenone ≥98.0% (by GC)

Supplier: TCI America

A2960-1G A2960-5G
TCA2960-1GEA 90 USD
TCA2960-1G TCA2960-5G
4'-Acetoxy-2'-hydroxyacetophenone ≥98.0% (by GC)
4'-Acetoxy-2'-hydroxyacetophenone
Formula: C₁₀H₁₀O₄
Boiling Pt: 303 °C
MDL Number: MFCD00092225
CAS Number: 42059-48-3

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 72.0 to 76.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR