1,2-Dibromo-2,4-dicyanobutane ≥98.0% (by GC)

Supplier: TCI America
Danger

B5387-25G B5387-5G
TCB5387-25GEA 77 USD
TCB5387-25G TCB5387-5G
1,2-Dibromo-2,4-dicyanobutane ≥98.0% (by GC)
1,2-Dibromo-2,4-dicyanobutane
Formula: C₆H₆Br₂N₂
Boiling Pt: 212 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00072483
CAS Number: 35691-65-7
EINECS: 252-681-0
UN: 3077
ADR: 9,III
Merck Index: 14,03021

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 53.0 to 58.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR