2'-Chloro-5'-nitroacetophenone ≥98.0% (by GC)

Supplier: TCI America
Warning

C3289-1G C3289-5G
TCC3289-1GEA 41 USD
TCC3289-1G TCC3289-5G
2'-Chloro-5'-nitroacetophenone ≥98.0% (by GC)
2'-Chloro-5'-nitroacetophenone
Formula: C₈H₆ClNO₃
MW: 199.5911 g/mol
MDL Number: MFCD00052836
CAS Number: 23082-50-0
EINECS: 245-421-2

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 60.0 to 64.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR