ω-Chloro-3,4-difluoroacetophenone ≥98.0% (by GC)

Supplier: TCI America
Danger

C3215-1G C3215-5G
TCC3215-1GEA 29 USD
TCC3215-1G TCC3215-5G
ω-Chloro-3,4-difluoroacetophenone ≥98.0% (by GC)
ω-Chloro-3,4-difluoroacetophenone
Formula: C₈H₅ClF₂O
MDL Number: MFCD03966888
CAS Number: 51336-95-9
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to Light yellow powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR