3-tert-Butylphenylboronic acid ≥98.0% (by HPLC, titration analysis) (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

B6001-1G B6001-5G
TCB6001-1GEA 41 USD
TCB6001-1G TCB6001-5G
3-tert-Butylphenylboronic acid ≥98.0% (by HPLC, titration analysis) (contains varying amounts of Anhydride)
3-tert-Butylphenylboronic acid
Formula: C₁₀H₁₅BO₂
MW: 178.0400 g/mol
Melting Pt: 221 °C
MDL Number: MFCD11110647
CAS Number: 560132-24-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) 98.0 to 110.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR