2-Bromo-4'-fluoro-3'-(trifluoromethyl)acetophenone ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 4-Fluoro-3-(trifluoromethyl)phenacyl bromide, 2-Bromo-1-(4-fluoro-3-(trifluoromethyl)phenyl)ethanone

B5213-1G B5213-5G
TCB5213-1GEA 69 USD
TCB5213-1G TCB5213-5G
2-Bromo-4'-fluoro-3'-(trifluoromethyl)acetophenone ≥98.0% (by GC)
2-Bromo-4'-fluoro-3'-(trifluoromethyl)acetophenone
Formula: C₉H₅BrF₄O
MW: 285.0330 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD03094509
CAS Number: 537050-14-9
UN: 3261
ADR: 8 III

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 42.0 to 46.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR