3-Carboxy-5-fluorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 3-Carboxy-5-fluorobenzeneboronic acid, 3-Borono-5-fluorobenzoic acid

C3287-1G C3287-5G
TCC3287-1GEA 100 USD
TCC3287-1G TCC3287-5G
3-Carboxy-5-fluorophenylboronic acid (contains varying amounts of Anhydride)
3-Carboxy-5-fluorophenylboronic acid
Formula: C₇H₆BFO₄
MW: 183.93 g/mol
MDL Number: MFCD07363791
CAS Number: 871329-84-9

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 95.0 to 111.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR