4-Chloro-(3-methoxycarbonyl)phenylboronic acid ≥97.0% (by titrimetric analysis) (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 4-Chloro-3-(methoxycarbonyl)benzeneboronic acid, 4-Chloro-3-(methoxycarbonyl)phenylboronic acid

C3320-1G C3320-200MG
TCC3320-1GEA 107 USD
TCC3320-1G TCC3320-200MG
4-Chloro-(3-methoxycarbonyl)phenylboronic acid ≥97.0% (by titrimetric analysis) (contains varying amounts of Anhydride)
4-Chloro-(3-methoxycarbonyl)phenylboronic acid
Formula: C₈H₈BClO₄
MW: 214.4107 g/mol
MDL Number: MFCD06801690
CAS Number: 874219-45-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 109.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR