3-Chloro-(4-methoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: (3-Chloro-4-(methoxycarbonyl)phenyl)boronic acid, 3-Chloro-4-(methoxycarbonyl)phenylboronic acid, 3-Chloro-4-(methoxycarbonyl)benzeneboronic acid

C3279-1G C3279-5G
TCC3279-1GEA 100 USD
TCC3279-1G TCC3279-5G
3-Chloro-(4-methoxycarbonyl)phenylboronic acid (contains varying amounts of Anhydride)
3-Chloro-(4-methoxycarbonyl)phenylboronic acid
Formula: C₈H₈BClO₄
MW: 214.4100 g/mol
MDL Number: MFCD06659874
CAS Number: 603122-82-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR