4-Carboxy-2-nitrophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 4-Carboxy-2-nitrobenzeneboronic acid

C3265-1G C3265-5G
TCC3265-1GEA 82 USD
TCC3265-1G TCC3265-5G
4-Carboxy-2-nitrophenylboronic acid (contains varying amounts of Anhydride)
4-Carboxy-2-nitrophenylboronic acid
Formula: C₇H₆BNO₆
MW: 210.94 g/mol
Melting Pt: 261 °C
MDL Number: MFCD00448780
CAS Number: 85107-54-6

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR