4-(Difluoromethoxy)benzonitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

D4955-1G D4955-5G
TCD4955-1GEA 43 USD
TCD4955-1G TCD4955-5G
4-(Difluoromethoxy)benzonitrile ≥98.0% (by GC)
4-(Difluoromethoxy)benzonitrile
Formula: C₈H₅F₂NO
MDL Number: MFCD00085006
CAS Number: 90446-25-6
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 40.0 to 44.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR