3-Bromo-4-iodoanisole ≥97.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2-Bromo-1-iodo-4-methoxybenzene

B5697-1G B5697-5G
TCB5697-1GEA 49 USD
TCB5697-1G TCB5697-5G
3-Bromo-4-iodoanisole ≥97.0% (by GC)
3-Bromo-4-iodoanisole
Formula: C₇H₆BrIO
MDL Number: MFCD12547848
CAS Number: 466639-53-2

Order Now

Specification Test Results

Appearance Colorless to Red to Green clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR