Diethylcyanamide ≥98.0% (by GC)

Supplier: TCI America
Danger

D4965-1G D4965-5G
TCD4965-1GEA 40 USD
TCD4965-1G TCD4965-5G
Diethylcyanamide ≥98.0% (by GC)
Diethylcyanamide
Formula: C₅H₁₀N₂
Boiling Pt: 188 °C
Flash Pt: 69 °C
MDL Number: MFCD00001769
CAS Number: 617-83-4
EINECS: 210-532-7
UN: 3276
ADR: 6.1,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR