Chloro[[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene](acetanilide)palladium(II)] ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

C2387-200MG C2387-1G
TCC2387-200MGEA 91 USD
TCC2387-200MG TCC2387-1G
Chloro[[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene](acetanilide)palladium(II)] ≥98.0% (by titrimetric analysis)
Chloro[[1,3-bis(2,6-diisopropylphenyl)imidazol-2-ylidene](acetanilide)palladium(II)]

This product is unavailable in the U.S.

Formula: C₃₅H₄₄ClN₃OPd
CAS Number: 1228009-69-5

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (Chelometric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR