2-Chloro-3-methyl-5-pyridineboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 2-Chloro-3-methylpyridine-5-boronic acid, 6-Chloro-5-methylpyridine-3-boronic acid, (6-Chloro-5-methylpyridin-3-yl)boronic acid

C3588-1G C3588-5G
TCC3588-1GEA 48 USD
TCC3588-1G TCC3588-5G
2-Chloro-3-methyl-5-pyridineboronic acid (contains varying amounts of Anhydride)
2-Chloro-3-methyl-5-pyridineboronic acid
Formula: C₆H₇BClNO₂
MW: 171.39 g/mol
MDL Number: MFCD03411564
CAS Number: 1003043-40-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 98.0 to 111.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR