2-(4-Chlorophenoxy)benzaldehyde ≥98.0% (by GC)

Supplier: TCI America

C3351-1G C3351-5G
TCC3351-1GEA 88 USD
TCC3351-1G TCC3351-5G
2-(4-Chlorophenoxy)benzaldehyde ≥98.0% (by GC)
2-(4-Chlorophenoxy)benzaldehyde
Formula: C₁₃H₉ClO₂
MW: 232.6600 g/mol
MDL Number: MFCD01568734
CAS Number: 111826-11-0

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 55.0 to 59.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR