6-Bromo-3-cyanochromone ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 6-Bromo-3-cyanochromone, 6-Bromo-4-oxo-4H-1-benzopyran-3-carbonitrile, 6-Bromochromone-3-carbonitrile, 6-Bromo-4-oxo-4H-chromene-3-carbonitrile

B4968-1G B4968-5G
TCB4968-1GEA 60 USD
TCB4968-1G TCB4968-5G
6-Bromo-3-cyanochromone ≥98.0% (by GC)
6-Bromo-3-cyanochromone
Formula: C₁₀H₄BrNO₂
MW: 250.483 g/mol
MDL Number: MFCD00191842
CAS Number: 52817-13-7
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 223.0 to 227.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR