2-(4-Bromophenyl)-5-phenyl-1,3,4-oxadiazole ≥97.0% (by HPLC)

Supplier: TCI America

B4513-1G B4513-5G
TCB4513-1GEA 90 USD
TCB4513-1G TCB4513-5G
2-(4-Bromophenyl)-5-phenyl-1,3,4-oxadiazole ≥97.0% (by HPLC)
2-(4-Bromophenyl)-5-phenyl-1,3,4-oxadiazole
Formula: C₁₄H₉BrN₂O
MW: 301.14 g/mol
MDL Number: MFCD00050997
CAS Number: 21510-43-0
EINECS: 244-413-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 97.0 area%
Purity (Argentmetric Titration) min. 95.0 %
Melting point 169.0 to 173.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR