4-Chloro-8-(trifluoromethyl)quinoline ≥98.0% (by GC)

Supplier: TCI America
Warning

C3067-1G C3067-5G
TCC3067-1GEA 109 USD
TCC3067-1G TCC3067-5G
4-Chloro-8-(trifluoromethyl)quinoline ≥98.0% (by GC)
4-Chloro-8-(trifluoromethyl)quinoline
Formula: C₁₀H₅ClF₃N
MW: 231.61 g/mol
MDL Number: MFCD00134578
CAS Number: 23779-97-7
EINECS: 245-880-9

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 78.0 to 82.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR