4-Chloro-2-(trifluoromethyl)quinoline ≥97.0% (by GC)

Supplier: TCI America
Warning

C3262-1G C3262-5G
TCC3262-1GEA 52 USD
TCC3262-1G TCC3262-5G
4-Chloro-2-(trifluoromethyl)quinoline ≥97.0% (by GC)
4-Chloro-2-(trifluoromethyl)quinoline
Formula: C₁₀H₅ClF₃N
MW: 231.61 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00153105
CAS Number: 1701-24-2

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Melting point 34.0 to 38.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR