α,α-Difluoroanisole ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: (Difluoromethoxy)benzene, alpha,alpha-Difluoroanisole

D4914-25G D4914-5G
TCD4914-25GEA 282 USD
TCD4914-25G TCD4914-5G
α,α-Difluoroanisole ≥98.0% (by GC)
α,α-Difluoroanisole
Formula: C₇H₆F₂O
MW: 144.1200 g/mol
Boiling Pt: 140 °C
MDL Number: MFCD00068184
CAS Number: 458-92-4
EINECS: 207-283-1
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR