(2-Aminophenyl)acetonitrile ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 2-(2-Aminophenyl)acetonitrile, 2-Aminobenzyl cyanide, (2-Aminophenyl)acetonitrile

A2997-1G A2997-200MG
TCA2997-1GEA 160 USD
TCA2997-1G TCA2997-200MG
(2-Aminophenyl)acetonitrile ≥98.0% (by GC, titration analysis)
(2-Aminophenyl)acetonitrile
Formula: C₈H₈N₂
MW: 132.17 g/mol
MDL Number: MFCD00091082
CAS Number: 2973-50-4
EINECS: 221-015-0
UN: 3439
ADR: 6.1,III

Order Now

Specification Test Results

Appearance White to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 69.0 to 73.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR