3-Coumaranone ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Benzofuran-3(2H)-one

C2825-1G C2825-5G
TCC2825-1GEA 45 USD
TCC2825-1G TCC2825-5G
3-Coumaranone ≥98.0% (by GC)
3-Coumaranone
Formula: C₈H₆O₂
MW: 134.13 g/mol
MDL Number: MFCD00051810
CAS Number: 7169-34-8

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 98.0 %
Melting point 98.0 to 102.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR