1-Bromo-3-fluoro-2-iodobenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

B5982-1G B5982-5G
TCB5982-1GEA 45 USD
TCB5982-1G TCB5982-5G
1-Bromo-3-fluoro-2-iodobenzene ≥98.0% (by GC)
1-Bromo-3-fluoro-2-iodobenzene
Formula: C₆H₃BrFI
Boiling Pt: 109 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07368675
CAS Number: 450412-29-0

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR