1-Bromo-2-chloro-4-iodobenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

B5742-1G B5742-5G
TCB5742-1GEA 19 USD
TCB5742-1G TCB5742-5G
1-Bromo-2-chloro-4-iodobenzene ≥98.0% (by GC)
1-Bromo-2-chloro-4-iodobenzene
Formula: C₆H₃BrCl
MDL Number: MFCD04973904
CAS Number: 535934-25-9

Order Now

Specification Test Results

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 38.0 to 42.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR