4-Chlorobenzo[b]thiophene-2-carboxylic acid ≥97.0% (by GC)
Supplier: TCI America
|
Warning
|
C3430-1G
C3430-5G
TCC3430-1GEA
89
USD
TCC3430-1G
TCC3430-5G
4-Chlorobenzo[b]thiophene-2-carboxylic acid ≥97.0% (by GC)
4-Chlorobenzo[b]thiophene-2-carboxylic acid
Formula:
C₉H₅ClO₂S MW: 212.65 g/mol |
MDL Number:
MFCD06655155 CAS Number: 23967-57-9 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 97.0 % |
Purity (Neutralization titration) | min. 96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...