1-(2-Chloroethoxy)-4-nitrobenzene ≥98.0% (by GC)

Supplier: TCI America
Warning

C3523-1G C3523-5G
TCC3523-1GEA 79 USD
TCC3523-1G TCC3523-5G
1-(2-Chloroethoxy)-4-nitrobenzene ≥98.0% (by GC)
1-(2-Chloroethoxy)-4-nitrobenzene
Formula: C₈H₈ClNO₃
MW: 201.6100 g/mol
Boiling Pt: 206 °C
MDL Number: MFCD00674505
CAS Number: 3383-72-0

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 57.0 to 61.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR